missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq DNA Polymerase (5U/μL)

Catalog No. 11EPTQL100
Change view
Click to view available options
Quantity:
1000U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11EPTQL100 1000U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11EPTQL100 Supplier MP Biomedicals™ Supplier No. 11EPTQL100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Taq DNA Polymerase, 5X Direct Loading Buffer with Mg, 25mM MgCl2

Includes Direct Loading Buffer for loading of sample onto agarose gel after amplification

Taq polymerase catalyzes oligonucleotide primer-driven, DNA template dependent incorporation of dNTPs into complimentary DNA strands. It displays both 5' to 3' polymerase and exonuclease activities. The included 5X Direct Loading buffer contains a densifying agent and a red-purple dye that allows the migration of the sample to be tracked during electrophoresis.

Unit Definition: One unit will incorporate 10nmol of total dNTPs into acid-precipitable DNA in 30 min. at 74°C.

Specifications

Concentration 5 U/μL
Content And Storage Store at -20°C
Quantity 1000U
For Use With (Application) Standard PCR, RT-PCR, Sequencing, Nucleic Acid Labeling
Includes Taq DNA Polymerase, 5X Direct Loading Buffer with Mg, 25mM MgCl2