missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TAPS, 99+%, for biochemistry
CAS: 29915-38-6 | C7H17NO6S | 243.27 g/mol
$74.49 - $181.59
Chemical Identifiers
| CAS | 29915-38-6 |
|---|---|
| Molecular Formula | C7H17NO6S |
| Molecular Weight (g/mol) | 243.27 |
| MDL Number | MFCD00007538 |
| InChI Key | YNLCVAQJIKOXER-UHFFFAOYSA-N |
| Synonym | 3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid |
| PubChem CID | 121591 |
| IUPAC Name | 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid |
| SMILES | OCC(CO)(CO)NCCCS(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC327800250
|
Thermo Scientific Chemicals
327800250 |
25 g | Glass bottle |
Each for $74.49
|
|
||||
|
AC327801000
|
Thermo Scientific Chemicals
327801000 |
100 g | Glass bottle |
Each for $181.59
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
| 29915-38-6 | |
| 243.27 | |
| YNLCVAQJIKOXER-UHFFFAOYSA-N | |
| 121591 | |
| OCC(CO)(CO)NCCCS(O)(=O)=O |
| C7H17NO6S | |
| MFCD00007538 | |
| 3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid | |
| 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid |
Specifications
| (0.1 M aq. soln.) | |
| 240.0°C | |
| White | |
| Powder | |
| >110°C | |
| Authentic | |
| (HOCH2)3CNH(CH2)3SO3H | |
| Glass bottle | |
| 3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid | |
| YNLCVAQJIKOXER-UHFFFAOYSA-N | |
| 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid | |
| 121591 | |
| ≥99% | |
| TAPS |
| (0.1M aq. soln.) | |
| 29915-38-6 | |
| 5 to 6.5 (1% aq. soln. at 25°C) | |
| 25 g | |
| 99+% | |
| C7H17NO6S | |
| MFCD00007538 | |
| 15, 9191 | |
| Solubility in water: ca 121.6g/L (20°C). | |
| OCC(CO)(CO)NCCCS(O)(=O)=O | |
| 243.27 | |
| 243.27 | |
| Biochemical |
RUO – Research Use Only