missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TAPS, 99+%, for biochemistry

CAS: 29915-38-6 | C7H17NO6S | 243.27 g/mol

Supplier:  Thermo Scientific Chemicals 327801000

 View more versions of this product

Catalog No. AC327801000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
Specifications

Specifications

0.04 max. at 260nm
(0.1M aq. soln.)
29915-38-6
5 to 6.5 (1% aq. soln. at 25°C)
100 g
99+%
C7H17NO6S
MFCD00007538
15, 9191
Solubility in water: ca 121.6g/L (20°C).
OCC(CO)(CO)NCCCS(O)(=O)=O
243.27
243.27
Biochemical
TAPS
240.0°C
White
Powder
>110°C
Authentic
(HOCH2)3CNH(CH2)3SO3H
Glass bottle
3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid
YNLCVAQJIKOXER-UHFFFAOYSA-N
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
121591
≥99%

Chemical Identifiers

29915-38-6
243.27
YNLCVAQJIKOXER-UHFFFAOYSA-N
121591
OCC(CO)(CO)NCCCS(O)(=O)=O
C7H17NO6S
MFCD00007538
3-[Tris(hydroxymethyl)methylamino]-1-propanesulfonic acid
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only