missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TAPS, 0.2M buffer soln., pH 8.0

Catalog No. AAJ60322AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60322AE 100 mL
AAJ60322AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60322AE Supplier Thermo Scientific Chemicals Supplier No. J60322AE

CAS: 91000-53-2265.256 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91000-53-2
Molecular Weight (g/mol) 265.256

Specifications

Concentration 0.2 M
pH 8.0
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material TAPS
Recommended Storage Ambient temperatures

RUO – Research Use Only