missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tangeretin 96.0+%, TCI America™
$35.41 - $201.32
Chemical Identifiers
| CAS | 481-53-8 |
|---|---|
| Molecular Formula | C20H20O7 |
| Molecular Weight (g/mol) | 372.37 |
| MDL Number | MFCD00017438 |
| InChI Key | ULSUXBXHSYSGDT-UHFFFAOYSA-N |
| PubChem CID | 68077 |
| ChEBI | CHEBI:9400 |
| IUPAC Name | 5,6,7,8-tetramethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one |
| SMILES | COC1=CC=C(C=C1)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T270810MG
|
TCI America
T270810MG |
10 mg |
Each for $35.41
|
|
|||||
|
T2708100MG
|
TCI America
T2708100MG |
100 mg |
Each for $201.32
|
|
|||||
Chemical Identifiers
| 481-53-8 | |
| 372.37 | |
| ULSUXBXHSYSGDT-UHFFFAOYSA-N | |
| CHEBI:9400 | |
| COC1=CC=C(C=C1)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1 |
| C20H20O7 | |
| MFCD00017438 | |
| 68077 | |
| 5,6,7,8-tetramethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one |
Specifications
| 481-53-8 | |
| White | |
| MFCD00017438 | |
| 4′C,5,6,7,8-Pentamethoxyflavone | |
| COC1=CC=C(C=C1)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1 | |
| 372.37 | |
| CHEBI:9400 | |
| ≥96.0% (HPLC) | |
| Tangeretin |
| 155°C | |
| C20H20O7 | |
| 10 mg | |
| ULSUXBXHSYSGDT-UHFFFAOYSA-N | |
| 5,6,7,8-tetramethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one | |
| 68077 | |
| 372.37 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DJ3102725
TSCA : No
Recommended Storage : Refrigerator