missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Tangeretin | |
| 155°C | |
| C20H20O7 | |
| 10 mg | |
| ULSUXBXHSYSGDT-UHFFFAOYSA-N | |
| 5,6,7,8-tetramethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one | |
| 68077 | |
| 372.37 | |
| Crystalline Powder |
| 481-53-8 | |
| White | |
| MFCD00017438 | |
| 4′C,5,6,7,8-Pentamethoxyflavone | |
| COC1=CC=C(C=C1)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1 | |
| 372.37 | |
| CHEBI:9400 | |
| ≥96.0% (HPLC) |
Chemical Identifiers
| 481-53-8 | |
| 372.37 | |
| ULSUXBXHSYSGDT-UHFFFAOYSA-N | |
| CHEBI:9400 | |
| COC1=CC=C(C=C1)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1 |
| C20H20O7 | |
| MFCD00017438 | |
| 68077 | |
| 5,6,7,8-tetramethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one |
Safety and Handling
RTECSNumber : DJ3102725
TSCA : No
Recommended Storage : Refrigerator