missing translation for 'onlineSavingsMsg'
Learn More

Tangeretin 96.0+%, TCI America™

Supplier:  TCI America T2708100MG

 View more versions of this product

Catalog No. T2708100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tangeretin
155°C
C20H20O7
100 mg
ULSUXBXHSYSGDT-UHFFFAOYSA-N
5,6,7,8-tetramethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one
68077
372.37
Crystalline Powder
481-53-8
White
MFCD00017438
4′,5,6,7,8-Pentamethoxyflavone
COC1=CC=C(C=C1)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1
372.37
CHEBI:9400
≥96.0% (HPLC)

Chemical Identifiers

481-53-8
372.37
ULSUXBXHSYSGDT-UHFFFAOYSA-N
CHEBI:9400
COC1=CC=C(C=C1)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1
C20H20O7
MFCD00017438
68077
5,6,7,8-tetramethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DJ3102725

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions