accessibility menu, dialog, popup

UserName

CAS RN 481-53-8

CH3OOOCH3OCH3OCH3OH3CO

CAS RN 481-53-8

IUPAC Name: 5,6,7,8-tetramethoxy-2-(4-methoxyphenyl)-4H-chromen-4-one
Synonyms: tangeretin ponkanetin pentamethoxyflavone tangeretin (6CI)
Molecular Weight (g/mol): 372.37
Molecular Formula: C20H20O7
InChi Key: ULSUXBXHSYSGDT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results