missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ TallPrep™ Metal Bead Lysing Matrix Tubes

TallPrep™ Metal Bead Lysing Matrix Tubes for nucleic acid isolation and purification

Supplier:  MP Biomedicals™ 116945050

 View more versions of this product

Catalog No. MP116945050


Only null left
Explore available promotions

Description

Description

  • Each impact-resistant 4.5 mL tube contains twelve 3.175 mm (1/8 in.) diameter stainless steel beads
  • Use to break hard/brittle samples
  • Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing
Specifications

Specifications

Stainless Steel
Store at Room Temperature.
DNase Free
Nucleic Acid isolation and purification
Lysis Matrix Tube
RNase Free
4.5 mL
Animal Tissue, Roots, Bone, Skeletal Muscle, Sludge, Food, Fungi, Wastewater, Lung, Bacteria (Gram +/-), Bone, Feces, Fixed Samples, Food, Fungi, Heart, Lung, Plants
FastPrep-24, FastPrep 120, FastPrep-96
Stainless Steel
50 x 4.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions