missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ TallPrep™ Metal Bead Lysing Matrix Tubes
TallPrep™ Metal Bead Lysing Matrix Tubes for nucleic acid isolation and purification
Supplier: MP Biomedicals™ 116945100
Description
- Each impact-resistant 4.5 mL tube contains twelve 3.175 mm (1/8 in.) diameter stainless steel beads
- Use to break hard/brittle samples
- Specialty applications include RNA and protein extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing
Specifications
| Stainless Steel | |
| Store at Room Temperature. | |
| DNase Free | |
| RNase Free |
| 4.5 mL | |
| Bone, Heart, Lung, Seeds, Skeletal Muscle, Spores, Food, Plants, Roots, Fixed Samples, Tough Tissues, All Samples | |
| 100 x 4.5 mL |