Learn More
Synthware™ Inlet Adapters with Fritted Disc
GREENER_CHOICE
With extra coarse porosity fritted disc to prevent solid from being introduced into reaction system during liquid transfer
$86.36 - $91.18
Spécifications
For Use With (Application) Organic Synthesis
Material Borosilicate Glass (3.3)
Product Type Inlet Adapter with Fritted Disc
Quantity 1
voir les spécifications
Produits 3
Numéro de catalogue Numéro du manufacturier. Standard Taper Joint No. Height (Metric) Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Standard Taper Joint No. Height (Metric) Prix Quantity & Availability  
31500545
GREENER_CHOICE
Afficher les documents Détails de la promotion
Synthware™
A831420
14/20 65mm
chaque for $86.36
Il en reste null
31500546
GREENER_CHOICE
Afficher les documents Détails de la promotion
Synthware™
A831922
19/22 67mm
chaque for $88.46
Il en reste null
31500547
GREENER_CHOICE
Afficher les documents Détails de la promotion
Synthware™
A832440
24/40 95mm
chaque for $91.18
Il en reste null
Description
Description
Synthware inlet adapter with an extra coarse porosity fritted disc between the serrated hose connection and the standard taper inner joint. The disc prevents solids from being introduced into the reaction system during liquid transfer.
Spécifications
Spécifications
Organic Synthesis
Borosilicate Glass (3.3)
Inlet Adapter with Fritted Disc
1
Videos
FDS