Learn More
Synthware™ Distilling Head
GREENER_CHOICE
Designed for use with a 76mm immersion thermometer
$386.71 - $424.75
Specifications
Type Distillation Head
Standard Taper Joint Thermometer Joint 10/18
Joint Type 24/40
View More Specs
Products 3
Catalog Number Mfr. No. Length (Metric) Jacket Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Jacket Price Quantity & Availability  
31501028
GREENER_CHOICE
View Documents Promotion Details
Synthware™
D304250
250mm
Each for $397.14
Only null left
31501027
GREENER_CHOICE
View Documents Promotion Details
Synthware™
D304150
150mm
Each for $386.71
Only null left
31501029
GREENER_CHOICE
View Documents Promotion Details
Synthware™
D304400
400mm
Each for $424.75
Only null left
Description
Description
  • Distillation head with condenser side arm sealed at 75 deg. angle to adapter
  • Inner joint on uptake tube and inner drip tip joint on condenser of same size
Specifications
Specifications
Distillation Head
Thermometer Joint 10/18
24/40