Learn More
Synthware™ Distilling Head
GREENER_CHOICE
Catalog No. 31501029
Change view
Click to view available options
Length (Metric) Jacket:
150mm
250mm
400mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Jacket
31501029 400mm
31501028 250mm
31501027 150mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31501029 Supplier Synthware™ Supplier No. D304400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed for use with a 76mm immersion thermometer
  • Distillation head with condenser side arm sealed at 75 deg. angle to adapter
  • Inner joint on uptake tube and inner drip tip joint on condenser of same size
Specifications
Type Distillation Head
Standard Taper Joint Thermometer Joint 10/18
Length (Metric) Jacket 400mm
Joint Type 24/40