Learn More
Synthware™ Distilling Head
GREENER_CHOICE
Designed for use with a 76mm immersion thermometer
Supplier:  Synthware™ D304150
 View more versions of this product
Catalog No. 31501027
Only null left
Description
Description
  • Distillation head with condenser side arm sealed at 75 deg. angle to adapter
  • Inner joint on uptake tube and inner drip tip joint on condenser of same size
Specifications
Chemical Identifiers
Distillation Head
Thermometer Joint 10/18
24/40
Specifications
Distillation Head
Thermometer Joint 10/18
150mm
24/40
Product Suggestions
Product Suggestions