Learn More
Synthware™ Air Reflux Condenser, Microscale
Description
- Supplied with one-holed Nylon™ compression capand one Viton™O-ring
Specifications
Specifications
| Type | Air Reflux Condenser |
| Material | Glass |
| Standard Taper Joint No | 14/10 |
| Length (Metric) | 100 mm |