missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Synthetic Laminin Peptide for Rat Neural Stem Cells

Catalog No. SCR127
Change view
Click to view available options
Quantity:
5 x 3 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCR127 5 x 3 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCR127 Supplier MilliporeSigma™ Supplier No. SCR127

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For cell culture applications

Specifications

For Use With (Application) Cell Culture
Quantity 5 x 3 mg
Product Type Laminin Peptide
Content And Storage -20°C
Form Lyophilized
Purity or Quality Grade >95% Purity by HPLC Analysis