Learn More
Agilent BioTek Synergy LX Multi-Mode Reader, IVD

The BioTek Synergy LX multi-mode reader automates many common microplate assays. For In Vitro Diagnostic Use.
Supplier: Agilent BioTek SLXFATSSI
Description
The BioTek Synergy LX multi-mode reader automates many common microplate assays. The high-quality optic design ensures excellent data in absorbance, fluorescence, and luminescence detection modes. This affordable system is ideal for many common assays including nucleic acid and protein quantification, ELISA, BCA, Bradford, and cell viability
The data display, output to a USB flash drive, printer, or Gen5 software make Synergy LX a versatile assay workstation. The large touch screen user interface and onboard software simplify programming and save time in operation of microplate and microvolume assays.
Identifiants chimiques
| 200 to 900 nm | |
| CE and TUV marked. RoHS compliant. | |
| 38.1 cm | |
| 24 sec. (96 wells), 76 sec. (384 wells) | |
| Monochromator Bandpass | |
| 50/60 Hz | |
| 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) | |
| Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP | |
| English | |
| 100/240 V | |
| 12.3 kg | |
| 38.1 cm | |
| Full data analysis and reporting (under Gen5 control) | |
| End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning |
| Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD | |
| ≤5 nm | |
| 15 in. | |
| Linear, Orbital, Double Orbital | |
| Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability | |
| <1% from 0 to 2.5 OD | |
| 0.001 OD | |
| 2 | |
| 1 year | |
| 27 lb. | |
| 15 in. | |
| Photodiode, PMT | |
| Xenon Flash |
Spécifications
| 200 to 900 nm | |
| CE and TUV marked. RoHS compliant. | |
| 38.1 cm | |
| 24 sec. (96 wells), 76 sec. (384 wells) | |
| Monochromator Bandpass | |
| 15 in. | |
| 38.1 cm | |
| Absorbance, Fluorescence, Luminescence, Touchscreen | |
| Multi-Mode Microplate Reader | |
| 0.001 OD | |
| 2 | |
| 1 year | |
| 27 lb. | |
| 15 in. | |
| Photodiode, PMT | |
| Xenon Flash |
| Wavelength: ±2 nm; Optical Density: <1% at 2.0 OD, <3% at 2.5 OD | |
| ≤5 nm | |
| 15 in. | |
| Linear, Orbital, Double Orbital | |
| Nucleic acid and protein quantification, ELISA, BCA, Bradford and cell viability | |
| 50/60 Hz | |
| <1% from 0 to 2.5 OD | |
| SLXFATS | |
| 24 VDC power supply compatible with 100/240 VAC at 50/60 Hz, 60 W (max.) | |
| Fluorescence Intensity: Fluorescein 2 pM; Luminesence: 10 amol ATP | |
| English | |
| 100/240 V | |
| 12.3 kg | |
| 38.1 cm | |
| Full data analysis and reporting (under Gen5 control) | |
| End Point, Kinetic, Area Scanning, Absorbance Spectral Scanning |