Learn More
Walter Products™ Suture Training Pad, 4 x 4 in.
Catalog No. S139039
Change view
Click to view available options
Dimensions (L x W):
4 x 4 in.
Catalog No. Dimensions (L x W)
S139039 4 x 4 in.
1 options
Catalog No. S139039 Supplier Walter Products™ Supplier No. SP1001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Reusable training pad for suturing technique.
  • Ideal for mastering suturing technique
  • Reusable
  • Simulates anatomical structure of human tissue including 3 layers (skin, fat and muscle)
  • 11 pre-cut wounds of various types of width, depth and lots of space to add additional cuts
  • Integrated mesh layer tolerates tension and whole suturing
  • Includes instruction manual
Specifications
Material Silicone
Dimensions (L x W) 4 x 4 in.
For Use With (Application) Suturing Practice
Includes Practice Pad