Learn More
FEATHER Surgical Blade (Microsurgery)
Description
- Made in Japan
- Individually packed
- Single use only
Spécifications
Spécifications
| Blade Material | Stainless Steel |
| Size | # 370 |
| For Use With (Equipment) | FEATHER Surgical Handle # 3, # 3L, # 7, or # DF-160S-3 |
| Disposable | Single-use |
| Sterility | Sterile |
| Quantity | 10/Pk. |