missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ SureOne™ Large Orifice Non-Filtered Pipette Tips
GREENER_CHOICE

Catalog No. 02707464
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Format:
Bulk
Hinged Rack
Hinged Rack Sterile
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Format
02707464 1 to 200 μL Sterile Hinged Rack Sterile
02707459 100 to 1000 μL Non-sterile Hinged Rack
02707460 100 to 1000 μL Sterile Hinged Rack Sterile
02707463 1 to 200 μL Non-sterile Hinged Rack
02707458 100 to 1000 μL Non-sterile Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02707464 Supplier Fisherbrand™ Supplier No. T118RSFIS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Using Fisherbrand™ Sure One™ large orifice pipette tips can help maintain the integrity of genomic DNA (gDNA) and reduce the risk of cell shearing during pipetting. With a 1.5 mm inner diameter tip orifice helps reduce shearing forces and are best suited for handling viscous or particulate solutions.

  • Large orifice, 1.5 mm inner diameter, helps reduce shearing forces
  • Low Retention, hydrophobic feature provides maximum sample recovery
  • Non-sterile products are certified to be free of RNase, DNase, DNA and Pyrogen
  • Sterilized products are certified to be free of RNase, DNase, DNA, Pyrogen/Endotoxin, PCR Inhibitors, ATP and Bioburden
  • Universal Fit, compatible with most popular brands of pipettor (optimal fit with Fisherbrand Elite™ pipettor)

Available bulk, rack non-sterile, rack sterile, and filtered rack sterile

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Length (English) 1.96 in.
Length (Metric) 4.97 cm
Clean Claims Sterilized products are certified to be free of RNase, DNase, DNA, Pyrogen/Endotoxin, PCR Inhibitors, ATP and Bioburden
Format Hinged Rack Sterile
Tip Style Genomic/Wide Orifice
Packaging 96 Tips/Rack, 960/Pk., 4800/Cs.
Product Type Non-Filtered Pipette Tip
For Use With (Equipment) Universal Fit, fits most pipettes
Graduated No
Material Polypropylene
Show More Show Less

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.