Learn More
Invitrogen™ SureLock™ Tandem Midi Gel Tank
Description
The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.
Features:
- 2-in-1 gel electrophoresis and transfer tank—enables electrophoresis and transfer of high-performance Invitrogen midi gels using the same tank
- Two separate chambers—run 1 or 2 gels or transfers and use only the necessary amount of buffer for each gel, minimizing buffer cost and waste
- Optimal performance with fast protocols—saves time while yielding better results
- Room temperature transfer when paired with SureLock Tandem Midi Blot Modules—eliminates the need to pre-chill buffers and the hassle and messiness of ice baths
- User-friendly—easy setup and simple workflow
- Does not require dummy cassette or buffer dam—reduces accessories
- Compatible—fits all Invitrogen precast midi gels and midi gel cassettes
- Modern design provides easy access to sample loading area reducing sample loading time and effort
- Robust design using strong polycarbonate plastic that prevents cracks and leaks
- Unobstructed visibility to your gels as they run
- Easy fitting lid with guiding notches and wire management hooks
The SureLock Tandem Midi Gel Tank can be used for wet tank transfers when paired with the SureLock Tandem Midi Blot Module. The SureLock Tandem Midi Blot Module performs efficient, room-temperature, wet protein transfers for downstream western blot analysis. The tank accommodates two blot modules, allowing transfer of one or two gels at a time. The unique design of the SureLock Tandem Blot Module uses considerably less transfer buffer (only ∼300 mL per transfer) than other wet transfer systems. This lower buffer requirement keeps the amount of methanol waste (a hazardous material) to a minimum.
Specifications
Specifications
| Content And Storage | Contains: • 1 SureLock Tandem Midi Gel Tank • 1 Gel Knife |
| Gel Compatibility | All Invitrogen precast midi gels, and Invitrogen empty midi gel cassettes |
| Gel Size | Midi |
| Product Line | SureLock |
| Type | Midi Gel Tank |
| For Use With | All Invitrogen Precast Midi Gels, and Invitrogen Empty Midi Gel Cassettes |
| Gel Dimensions | Gel size: 8 x 13 cm (Midi or Wide Format Gels)Gel cassette: 10.3 x 15 cmThickness: 1.0 mm |
| Quantity | 1 tank |
| Shipping Condition | Room Temperature |
| Warranty | One year |
| Show More |
Frequently Asked Questions (FAQs)
If using a Bio-Rad™ power supply, Novex Power Supply Adapters (Cat. No. ZA10001) are NOT needed for the SureLock Tandem Midi Gel Tank and should be removed prior to use.
Unfortunately, the PowerEase Touch 120W power supply has a maximum output of 500 mA and is not suitable for transferring two midi gels. For wet transfer, we recommend using the PowerEase Touch Power Supply 350W or 600W unit, which can deliver up to 3 A. More information about these power supply units can be found on our website.
Yes, up to two midi gels can be run in the SureLock Tandem Midi Gel Tank using the PowerEase Touch 120W Power Supply.
The SureLock Tandem Midi Blot Module (Cat. No. STM2001) is compatible with the SureLock Tandem Midi Gel Tank.
Up to two midi gels can be run at a time using the SureLock Tandem Midi Gel Tank, each in separate chambers.