missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Gel Handcast Bundle A - Hardware and Reagents

Catalog No. HC1000SR
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HC1000SR 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HC1000SR Supplier Invitrogen™ Supplier No. HC1000SR
Save Up to 22% on Invitrogen Western Workflow Devices
View all promotions

Includes

The SureCast Handcast Bundle A includes all the hardware and reagents you need to get started:

  • 2 SureCast Handcast Stations
  • 2 SureCast™ Sealing Pads
  • 2 SureCast™ Glass Plate Sets (2 front and 2 back)
  • 6 SureCast™ Gel Combs (2 each: 10-, 12-, 15-well)
  • 10 SureCast™ Gel Spacers
  • 1 SureCast™ Stacking Buffer (1 L), 2-pack
  • 1 SureCast™ Resolving Buffer (1 L), 2-pack
  • 1 SureCast™ Acrylamide Solution (40%) (450 mL)
  • 1 SureCast™ APS (25 g)
  • 1 SureCast™ TEMED (30 mL)

Pour your own polyacrylamide gels easily and confidently with a SureCast™ Gel Handcast Bundle A. The SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

Innovate, leak-free design—minimizes the number of failed gels, resulting in less wasted time

  • Superior glass plate durability
  • Simple assembly of casting components—just drop the glass plates in, then lock the cam handle
  • Unique tilt feature for easy loading of acrylamide solutions

For a bundle that does not include the SureCast Handcast Reagents, please see SureCast™ Gel Handcast Bundle B. The system features durable glass plates that are securely sealed using a single-motion, load-and-lock mechanism. The unique tilt feature of the SureCast™ Handcast Station minimizes spillage and enhances loadability of casting solutions during handcasting. SureCast gels are designed for use in the Mini Gel Tank.

Specifications

Content And Storage • 2 SureCast Gel Handcast Stations
• 2 sets SureCast Glass Plates
• 10 SureCast Gel Spacers
• 1 SureCast 10-well Multi-Use Tool
• 1 SureCast 12-well Multi-Use Tool
• 1 SureCast 15-well Multi-Use Tool
• 1 set SureCast Gel Combs (10-, 12-, 15-well)
• 2 x 500 mL SureCast Stacking Buffer packs.

Store at room temperature.
• 5 x 500 mL SureCast Stacking Buffer packs.

Store at room temperature.
• 2 x 500 mL SureCast Resolving Buffer packs.

Store at room temperature.
• 5 x 500 mL SureCast Resolving Buffer packs.

Store at room temperature.
• 25 g SureCast Ammonium Persulfate.

Store at room temperature.
• 450 mL SureCast Acrylamade Solution.

Store at room temperature.
• 30 mL SureCast TEMED.

Store at room temperature.
Gel Size Mini
Product Line SureCast
Type Gel Handcast Bundle A
Quantity 1 kit
Shipping Condition Room Temperature
Do you offer the SureCast casting reagents separately?

The SureCast Handcast System and reagents are available both as a package or as standalones.

Can I reuse SureCast Glass Plates?

Yes, SureCastGlass Plates can be reused.They are up to 22 times more durable than Bio-Rad glass plates.

Why should I store all SureCast reagents at room temperature

Having everything at room temperature means not having to thaw reagents, thus saving time in gel casting.

For Research Use Only. Not for use in diagnostic procedures.