missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SureCast™ Multi-Use Tool, 12-well

Catalog No. HC1012
Encompass
Change view
Click to view available options
Quantity:
1 unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HC1012 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HC1012 Supplier Invitrogen™ Supplier No. HC1012

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These components of the SureCast Handcast System can be used to create polyacrylamide gels using SureCast Handcast Reagents as well as any other popular polyacrylamide gel casting reagents.

  • Pour polyacrylamide gels easily and confidently
  • For use with the SureCast vertical gel system
  • SureCast gels are designed for use in the Mini Gel Tank

Recommended Storage

Room Temperature

Warranty

Up to 12 months

Specifications

For Use With (Equipment) SureCast Vertical Gel System
Gel Size Mini
Product Line SureCast
Type 12-well Multi-Use Tool
Quantity 1 unit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.