missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GVS Supported Cellulose Acetate Membranes
Pure cellulose acetate impregnated on nonwoven support
Supplier: GVS 1214411
Description
- Exceptionally strong, Triton™-free, low protein-binding cellulose acetate impregnated onto nonwoven support
- Hydrophilic with unequaled dimensional and wet strength — eliminate filter cracking and tearing under pressure
- Fast flow, high throughput, low protein/nucleic acid binding
- Withstand temperatures to 180°C (356°F); autoclavable
- Mean thickness: 150μm
- White with plain surface; come in dispenser packs of 25 or 100
Protein and enzyme filtrations, tissue culture media preparation, biological/pharmaceutical solution filtration, sterility tests.
- In mL/min./cm2, measured at 520mmHg (10psi), 20°C (68°F).
- Pressure at which air is first forced through the pores of a water-wet membrane.
- Manufactured by GVS Maine
Specifications
| Autoclavable | |
| White | |
| 5 μm | |
| Protein and enzyme filtrations; tissue culture media preparation; biological/pharmaceutical solution filtration; sterility tests. | |
| 150 μm |
| 90psi | |
| Cellulose Acetate | |
| 375.0mL/min./cm sq. | |
| Hydrophilic | |
| 47 mm |