missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GVS Supported Cellulose Acetate Membranes
Description
- Exceptionally strong, Triton™-free, low protein-binding cellulose acetate impregnated onto nonwoven support
- Hydrophilic with unequaled dimensional and wet strength — eliminate filter cracking and tearing under pressure
- Fast flow, high throughput, low protein/nucleic acid binding
- Withstand temperatures to 180°C (356°F); autoclavable
- Mean thickness: 150μm
- White with plain surface; come in dispenser packs of 25 or 100
Protein and enzyme filtrations, tissue culture media preparation, biological/pharmaceutical solution filtration, sterility tests.
- In mL/min./cm2, measured at 520mmHg (10psi), 20°C (68°F).
- Pressure at which air is first forced through the pores of a water-wet membrane.
- Manufactured by GVS Maine
Specifications
Specifications
| Autoclavable | Autoclavable |
| Bubble Point | 50psi |
| Color | White |
| Material | Cellulose Acetate |
| Pore Size | 0.2 μm |
| Flow Rate | 20.0mL/min./cm sq. |
| For Use With (Application) | Protein and enzyme filtrations; tissue culture media preparation; biological/pharmaceutical solution filtration; sterility tests. |
| Hydrophobicity | Hydrophilic |
| Thickness (Metric) | 150 μm |
| Diameter (Metric) | 25 mm |