missing translation for 'onlineSavingsMsg'
Learn More

GVS Supported Cellulose Acetate Membranes

Pure cellulose acetate impregnated on nonwoven support

Supplier:  GVS 1212374

 View more versions of this product

Catalog No. A02SP01300


Only null left
Explore available promotions

Description

Description

  • Exceptionally strong, Triton™-free, low protein-binding cellulose acetate impregnated onto nonwoven support
  • Hydrophilic with unequaled dimensional and wet strength — eliminate filter cracking and tearing under pressure
  • Fast flow, high throughput, low protein/nucleic acid binding
  • Withstand temperatures to 180°C (356°F); autoclavable
  • Mean thickness: 150μm
  • White with plain surface; come in dispenser packs of 25 or 100

Protein and enzyme filtrations, tissue culture media preparation, biological/pharmaceutical solution filtration, sterility tests.

  1. In mL/min./cm2, measured at 520mmHg (10psi), 20°C (68°F).
  2. Pressure at which air is first forced through the pores of a water-wet membrane.
  3. Manufactured by GVS Maine

Specifications

Specifications

Autoclavable
White
0.2 μm
Protein and enzyme filtrations; tissue culture media preparation; biological/pharmaceutical solution filtration; sterility tests.
150 μm
50psi
Cellulose Acetate
20.0mL/min./cm sq.
Hydrophilic
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions