missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SuperSignal™ West Pico Complete Mouse IgG Detection Kit

Catalog No. PI34081
Change view
Click to view available options
Quantity:
1.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI34081 1.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI34081 Supplier Thermo Scientific™ Supplier No. LSG34081

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Combines SuperSignal West Pico substrate with HRP-conjugated rabbit or mouse secondary antibodies for a convenient, easy-to-use Western blot detection kit.

Thermo Scientific™ SuperSignal Western Blotting Kits are ideal for busy researchers or people new to Western Blotting.

All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate and are available for the detection of mouse and rabbit secondary primary antibodies. All SuperSignal Western Blotting kits come with complete, detailed instructions for those new to the technique of Western Blotting and abbreviated protocols for the experienced user. The kits are also available in a standard format (substrate and optimized HRP-conjugates) or a complete format which also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.

Highlights:

  • Standard and complete kits – choose kits with just secondary antibody and chemiluminescent substrate or kits that also include blocking buffer and wash buffer

  • Mouse and rabbit kits – choose kits with goat anti-mouse IgG or goat anti-rabbit IgG secondary antibody conjugates

  • Instructions – complete, detailed manual with recommended antibody dilutions

Ordering Information:

Each kit provides sufficient reagents to process 50 miniblots or 4000cm2 of membrane.

Includes:

Standard Detection Kits Include: West Pico Substrate (luminol/enhancer solution and stable peroxide solution), HRP-conjugated anti-rabbit IgG, anti-mouse IgG, or biotin-binding protein. Complete Detection Kits also include SuperBlock Blocking Buffer and TBS wash buffer.

Specifications

Description SuperSignal West Pico Complete Mouse IgG Detection Kit
Product Type Mouse IgG Detection Kit
Detection Method Chemiluminescence
For Use With (Application) Western Blot
Membrane Compatibility Nitrocellulose, PVDF
Product Line SuperSignal
Quantity 1.5 L
Conjugate HRP
Substrate Horseradish Peroxidase
Reactivity Mouse
Sufficient For 50 mini blots (4000 sq. cm of membrane)
Content And Storage Sufficient For: ∼62 mini blots probed with mouse primary antibodies
• SuperSignal West Pico Luminol/Enhancer Solution, 250 mL
• SuperSignal West Pico Stable Peroxide Solution, 250 mL
• Peroxidase Conjugated Goat anti-Mouse IgG (H + L), 2 mL
• SuperBlock Blocking Buffer, 1 L
• TBS Packs (each makes 500 mL), 4 packs
Show More Show Less
How can I increase the sensitivity of my Western blot?

Consider transferring to a different membrane or using a different detection method. We have observed increased sensitivity when using PVDF membranes in place of nitrocellulose. On PVDF membranes, using as little as 1 µg of total rat brain protein, PKC can be detected with alkaline phosphatase-mediated chromogenic detection in some cases using affinity-purified antibodies at a concentration of 0.5 µg/mL. Detection sensitivity can also be increased by using chemiluminescent detection, especially when using a SuperSignal West enhanced chemiluminescence subtrate (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/western-blotting/detect-proteins-western-blot/western-blot-detection-reagents/detection-technologies-western-blotting/chemiluminescent-western-blot-detection/supersignal-chemiluminescent-substrates.html) such as SuperSignal West Pico PLUS, SuperSignal West Dura, or SuperSignal West Femto. The secondary antibody should be used as recommend by the manufacturer and optimized as needed.

For Research Use Only. Not for use in diagnostic procedures.