missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Superose™ Media Prep Grade Lab Packs

Catalog No. 45000086
Change view
Click to view available options
Product Type:
Superose 12 Prep Grade
Superose 6 Prep Grade
Max. Pressure:
100 psi
58 psi
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Max. Pressure
45000086 Superose 6 Prep Grade 58 psi
45000093 Superose 12 Prep Grade 100 psi
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000086 Supplier Cytiva Supplier No. 17048901

High performance gel filtration media for scaleup to preparative purifications of biomolecules

  • Permit high recovery of a wide variety of biomolecules—proteins, peptides, polynucleotides, etc.
  • Supplied as bulk filtration media, for preparative separations
  • Intended as a complement to Superose PC prepacked columns; can be efficiently packed under medium pressure
  • Superose 6 Prep Grade intended for biomolecules of a wide fractionation range; Superose 12, for higher-molecular weight proteins

Order Info

Supplied in 125mL containers

Specifications

Quantity Each
Flow Rate 40 cm/hr.
Particle Size 30 to 40 μm
For Use With (Equipment) AKTA™ design pump or system
Bed Volume ≈100 mL
Product Type Superose 6 Prep Grade
For Use With (Application) Separations of Proteins and other biomolecules according to size
Max. Pressure 58 psi
pH Range 3 to 12
Support Type Highly Cross-linked Agarose
Sample Type Proteins, Peptides, Polynucleotides, Wide Variety of Biomolecules
Content And Storage 4 to 30°C, 20% Ethanol
Show More Show Less