Learn More
Epredia™ SuperFrost™ Plus Slides
Description
- Electrostatically attract tissue sections and cytology preparations, binding them to the slide
- Eliminate the need for special adhesives or protein coatings
- No background staining
- Available in a variety of colors
Specifications
Specifications
| Type | Microscope Slide |
| Material | Glass |
| Frosted | Yes |
| Color | White |
| Quantity | 144/Cs. |