missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Superdex™ 75 Prep Grade

Catalog No. 45002037
Change view
Click to view available options
Quantity:
150 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002037 150 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002037 Supplier Cytiva Supplier No. 17104401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offers high resolution fractionation for separation, polishing and product formulation of recombinant proteins

  • Gel filtration medium
  • For purifying proteins of fractionation range: 3000 to 70000Da
  • Average particle size: 34μm
  • Matrix: composite of cross-linked agarose and dextran
  • pH stability working range
  • Available in 150mL bottle

Specifications

Quantity 150 mL
Flow Rate 10 to 50 cm/hr.
Particle Size 24 to 44 μm
Product Type Superdex Media
Format Solid Spherical Bead
For Use With (Application) Separation, polishing and product formulation of recombinant Proteins
Max. Pressure 70psi
pH Range 3 to 12
Support Type Composite of cross-linked Agarose and Dextran
Sample Type Recombinant Protein
Content And Storage 4 to 30°C, 0.2 M sodium acetate in 20% Ethanol
Show More Show Less