missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Superdex 30 Increase Precision 3.2/300 GL Prepacked SEC Column

Catalog No. 45001001
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001001 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45001001 Supplier Cytiva Supplier No. 29219758

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for high resolution in small-scale preparative purification and analysis for sample volumes up to 500μL

  • Replacement for Superdex peptide columns
  • Reduced run time with maintained resolution for time saving or increased throughput
  • Higher resolution for improved protein purity and analysis data
  • Higher lot-to-lot consistency for

Specifications

Quantity Each
Flow Rate 0.075 mL/min1
Particle Size ∼9 μm1
Bed Volume 2.4mL
Product Type Superdex Pre-packed Column
Diameter (Metric) Inner 3.2mm
For Use With (Application) High resolution, small-scale purification and analysis of small biomolecules with Mr ˜ 100 to ˜7000, such as peptides
Material Column tube: Borosilicate Glass; Bed Support: Titatium
pH Range 3 to 12
Support Type Frit
Packaging Superdex Increase
Content And Storage 4°C to 30°C, 20% ethanol
Show More Show Less