missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Super-Q™ Plus High-Volume Water Purification System

Catalog No. ZFSQ115P4
Change view
Click to view available options
Type:
With Pump
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
ZFSQ115P4 With Pump
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZFSQ115P4 Supplier MilliporeSigma™ Supplier No. ZFSQ115P4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

Required filter cartridges each sold separately

  • Ion-Ex™ cartridge removes inorganics (CPMB 022 02)
  • Organex-Q™ cartridge removes trace organic contaminants (CPEX 02201)
  • For applications requiring particulate prefiltration and/or ultralow bacteria levels, contact your Fisher Scientific Customer Service Representative

With a Millipore Super-Q™ Plus High-Volume Water Purification System, you can easily produce up to 12L/min. of Type I ultrapure water

High-flow water systems provide final polish to water which has been pretreated by reverse osmosis, deionization or distillation. Eliminates need for water storage systems. Consistenlty produce ultrapure quality water that exceeds Type I ASTM™/CAP/CLSi specifications.

 

  • Water in bowls is automatically recirculated hourly to maintain water quality between uses
  • Gauge indicates pressure of incoming feedwater Display shows resistivity measurement Modular design allows customization for specific applications Also available with 1/3hp pump motor to move gravity-fed feedwater at a controlled flowrate and equalize flowrate in pressurized loops

 

Applications:

  • Produce water ideal for preparing tissue culture media and reagents, and as rinse water for electronics.

Order Info

Systems shown here are 4-bowl systems; 3-bowl system without pump (ZFSQ 115 03) is available through your Fisher Scientific Customer Service Representative.

Specifications

Contaminant Rejection (Microbiological) Less than10cfu/mL
Daily Output >20 L
Flow Rate 10 to 12
Product Water Type Type 1
Resistivity 18.2
Total Organic Carbon <20
Type With Pump
Model Super-Q Plus (4-bowl)
Dimensions (L x W x H) 8.75 x 45.25 x 31.5 in. (22 x 94.5 x 80 cm)
Length (English) 8.75 in.
Length (Metric) 22 cm
Width (English) 45.25 in.
Width (Metric) 94.5 cm
Height (English) 31.5 in.
Height (Metric) 80 cm
Depth (English) 8.7 in.
Depth (Metric) 22 cm
Weight (English) 140 lb.
Weight (Metric) 63.5 kg
Voltage 120 V
Show More Show Less