missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SUPELCOSIL™ ABZ+Plus Supelguard Cartridge, 5 μm Particle Size

Offer both high deactivation and unique selectivity.

$557.03 - $557.03

Specifications

Quantity 2/Pk.
Particle Size 5 μm
Length (Metric) 2 cm
Pore Size 120 Å
Type Guard Cartridge
View More Specs

Products 2
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Price Quantity  
111003883
View Documents
MilliporeSigma™ Supelco™
59605
2.1 mm
Each for $557.03
Only null left
 
111003880
View Documents
MilliporeSigma™ Supelco™
59535U
4 mm
Each for $557.03
Only null left
 
Description

Description

SUPELCOSIL™ ABZ+Plus columns offer both high deactivation and unique selectivity. Deactivated silica particles of very narrow particle size distribution ensure high efficiency with low back pressure. After bonding and endcapping reactions, the ABZ+Plus phase effectively shields unreacted silanol groups on the silica, preventing them from interacting with most analytes, and provides symmetric peaks regardless of an analyte's functionality. The phase also allows you to use low ionic strength buffers without having to add an ion-suppressing modifier. ABZ+Plus enables you to use simple mobile phases when analyzing the most difficult compounds; acids, strongly basic compounds, and zwitterions. Suitable for L60 per USP.
Specifications

Specifications

2/Pk.
2 cm
Guard Cartridge
HPLC
5 μm
120 Å
Reversed Phase
SUPELCOSIL Supelguard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC