missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SUPELCOSIL™ ABZ+Plus Supelguard Cartridge, 5 μm Particle Size
Offer both high deactivation and unique selectivity.
Supplier: MilliporeSigma™ Supelco™ 59535U
Description
SUPELCOSIL™ ABZ+Plus columns offer both high deactivation and unique selectivity. Deactivated silica particles of very narrow particle size distribution ensure high efficiency with low back pressure. After bonding and endcapping reactions, the ABZ+Plus phase effectively shields unreacted silanol groups on the silica, preventing them from interacting with most analytes, and provides symmetric peaks regardless of an analyte's functionality. The phase also allows you to use low ionic strength buffers without having to add an ion-suppressing modifier. ABZ+Plus enables you to use simple mobile phases when analyzing the most difficult compounds; acids, strongly basic compounds, and zwitterions. Suitable for L60 per USP.Specifications
| 2/Pk. | |
| 2 cm | |
| Guard Cartridge | |
| Reversed Phase | |
| SUPELCOSIL Supelguard |
| 5 μm | |
| 120 Å | |
| 4 mm | |
| HPLC |
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC