missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SUPELCOGEL™ Pb, 6% Crosslinked HPLC Column, 9 μm Particle Size
SUPELCOGEL™ Pb, 6% crosslinked HPLC column may be used for the separation and determination of d-mannose in human serum samples using liquid chromatography coupled with tandem mass spectrometry.
$2197.15 - $3191.69
Specifications
| Crosslinking | 0.06 |
|---|---|
| Diameter (Metric) | 7.8 mm |
| Particle Size | 9 μm |
| Stationary Phase | Sulfonated Polystyrene |
| Product Line | SUPELCOGEL |
Description
SUPELCOGEL™ Pb, 6% Crosslinked HPLC Column has been used for the separation of:. D-mannose in human serum samples during its determination using liquid chromatography-tandem mass spectrometry (LC-MS/MS) equipped with electrospray ionization (ESI) source and selected reaction monitoring (SRM) detection. Free sugars in switchgrass extractives during their quantification using high performance liquid chromatography equipped with refractive index (HPLC-RI) detector.. Total hemicellulose and cellulose sugars in sugarcane straw hydrolyzate during their determination using HPLC equipped with infra-red (IR) detector.Specifications
| 0.06 | |
| 9 μm | |
| SUPELCOGEL | |
| Ion Exclusion |
| 7.8 mm | |
| Sulfonated Polystyrene | |
| Sulfonated Polystyrene/Divinylbenzene Phase/Spherical Particle Platform | |
| Crosslinked HPLC |
SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC