missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SUPELCOGEL™ Pb, 6% Crosslinked HPLC Column, 9 μm Particle Size

Catalog No. 111003107
Change view
Click to view available options
Length (Metric):
10 cm
30 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111003107 10 cm
111003025 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111003107 Supplier MilliporeSigma™ Supelco™ Supplier No. 59335U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SUPELCOGEL™ Pb, 6% crosslinked HPLC column may be used for the separation and determination of d-mannose in human serum samples using liquid chromatography coupled with tandem mass spectrometry.

SUPELCOGEL™ Pb, 6% Crosslinked HPLC Column has been used for the separation of:. D-mannose in human serum samples during its determination using liquid chromatography-tandem mass spectrometry (LC-MS/MS) equipped with electrospray ionization (ESI) source and selected reaction monitoring (SRM) detection. Free sugars in switchgrass extractives during their quantification using high performance liquid chromatography equipped with refractive index (HPLC-RI) detector.. Total hemicellulose and cellulose sugars in sugarcane straw hydrolyzate during their determination using HPLC equipped with infra-red (IR) detector.

Specifications

Crosslinking 0.06
Diameter (Metric) 7.8 mm
Length (Metric) 10 cm
Particle Size 9 μm
Stationary Phase Sulfonated Polystyrene
Product Line SUPELCOGEL
Matrix Sulfonated Polystyrene/Divinylbenzene Phase/Spherical Particle Platform
Phase Ion Exclusion
Material Stainless Steel
Technique Crosslinked HPLC

SUPELCOGEL is a trademark of Sigma-Aldrich Co. LLC