Learn More
Thermo Scientific™ SUN-SRi™ Crimp Caps
Catalog No. 22030214
Change view
Click to view available options
Color:
Silver
Quantity:
500/Pk.
Material (Liner):
Without Septum
Catalog No. Color Quantity Material (Liner)
22030214 Silver 500/Pk. Without Septum
1 options
Catalog No. 22030214 Supplier Thermo Scientific™ Supplier No. 504 368
Only null left
Thermo Scientific caps and vials are available in a wide variety of sizes and configurations to fit the needs of many applications.
  • Tin plate caps or two-part composite tin plate and aluminum cap with or without septum
  • Two-part caps feature aluminum skirts that make the closures easier to crimp and remove
  • 20mm Tin Plate
Specifications
Material (Liner) Without Septum
Color Silver
Material Aluminum
Quantity 500/Pk.
Closure Size 20 mm
Product Type Crimp Cap