missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Sun, Earth and Moon Orbital Model

Catalog No. S12895
Change view
Click to view available options
Height (English):
13 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English)
S12895 13 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12895 Supplier Eisco™ Supplier No. BD0075

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Multiple moving axis on this gear driven Sun, Earth and Moon Orbital Model demonstrate daylight, night, seasons and phases of moon.

  • Mechanical gear-driven model
  • Multiple moving axis to demonstrate daylight, night, seasons and phases of the moon
  • 12 V light bulb inside sun demonstrates sunlight on moon and earth
  • Base labeled with the months of the year and seasons for each hemisphere
  • Requires 2 AA batteries, not included

Specifications

Height (Metric) 330 mm
Height (English) 13 in.
Product Type Sun, Earth and Moon Model