missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid (NF/FCC), Fisher Chemical

Catalog No. A484212
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A484212 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A484212 Supplier Thermo Fisher Scientific Formerly Fairlawn Supplier No. FLA484212
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3
Color Colorless to Brown
Melting Point 10°C
Boiling Point 290°C
Density 1.84g/cm³
Heavy Metals (as Pb) 5ppm max.
Identification Pass Test (FCC, NF), Pass Test (FCC)
Packaging Glass Bottle
Reducing Substances Pass Test (NF), Pass Test (FCC)
Residue after Ignition ≤0.005% (NF)
Vapor Pressure <0.001 mmHg at 20°C
Assay Percent Range 95.0 to 98.0 %
Quantity 2.5 L
Physical Form Liquid
UN Number 1830
Formula Weight 98.08
Percent Purity 95.0 to 98.0% (FCC)
Grade FCC/NF
Chemical Name or Material Sulfuric Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
DOTInformation Class 8: Corrosive