missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Fuming, 18-24%, Spectrum™ Chemical

Catalog No. 18614287
Change view
Click to view available options
Quantity:
9 lb.
Packaging:
Poly-Coated Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614287 9 lb. Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18614287 Supplier Spectrum Chemical Mfg Cor Supplier No. S14239LB
Only null left

Chemical Identifiers

CAS 8014-95-7 , 7446-11-9

Specifications

CAS Max % 1
Packaging Poly-Coated Glass Bottle
Quantity 9 lb.
Percent Purity 18 to 24%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.