missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, For Phosphorus, Certified, 14% (v/v) ±1%, LabChem™
Specifications
Specifications
| Color | Colorless |
| Density | 1.15g/mL |
| CAS Max % | 78.52, 21.48 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1.15g/mL |
| Traceability to NIST | Traceable to NIST |
| Decomposition Information | Sulfur compounds; Thermal decomposition generates: Corrosive vapors |
| Quantity | 1 L |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves/eye protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/docto |
| Recommended Storage | Room Temperature |