missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Concentrate 0.1 M H2SO4 in Water (0.2 N), Eluent concentrate for IC, MilliporeSigma™ Supelco™

Catalog No. 111020292
Change view
Click to view available options
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111020292 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020292 Supplier Merck Emd Millipore Supplier No. 682791L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Prepared with sulfuric acid and high purity water (18.2 MOmega, 0.2 μm filtered)

Specifications

Chemical Name or Material Sulfuric Acid
CAS 7664-93-9
Quantity 1 L
Linear Formula H2SO4
MDL Number MFCD00064589
Synonym Sulfuric acid solution
Molecular Weight (g/mol) 98.08
Formula Weight 98.08