missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Babcock, Macron Fine Chemicals™
macron
Supplier: Avantor H37807
Specifications
| Sulfuric acid | |
| 1.84g/cm3 | |
| H2O4S | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.079 |
| 7664-93-9 | |
| 4L | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(=O)(=O)O | |
| 98.072 | |
| CHEBI:26836 | |
| FCC |
Chemical Identifiers
| 7664-93-9 | |
| 98.072 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(=O)(=O)O |
| H2O4S | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |