missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, for analysis, 25% solution in water

Catalog No. AC451050010
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451050010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC451050010 Supplier Thermo Scientific Chemicals Supplier No. 451050010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Heavy Metals (as Pb) 1ppm max.
Residue after Ignition 0.0005% max.
Linear Formula H2SO4
Quantity 1 L
Solubility Information Solubility in water: soluble.
Formula Weight 98.08
Grade Analytical
Chemical Name or Material Sulfuric acid
Hazard Category Danger
Hazard Statement GHS H Statement Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Do not breathe dust/fume/gas/mist/vapors/spray.

RUO – Research Use Only