missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 9.00 Normal, Reagents

Catalog No. 01333708
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01333708 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01333708 Supplier Labchem Inc Supplier No. CS1343001A
Only null left

Sulfuric Acid, 9.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH 0
Color Colorless
Boiling Point 107°C
Packaging Poly Bottle
Specific Gravity 1.26
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 9.00 Normal