missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 85% (v/v), Ricca Chemical

Catalog No. 8190851
Change view
Click to view available options
Quantity:
4 L
Packaging:
Safety-coated Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8190851 4 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 8190851 Supplier Ricca Chemical Company Supplier No. 8190.851
Only null left

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 85% (v/v)
CAS Min % 84.0, 14.0
CAS Max % 86.0, 16.0
Packaging Safety-coated Glass Bottle
Solvent or Matrix Aqueous Solution
Quantity 4 L
Physical Form Liquid
Percent Purity 85% V/V
Grade Laboratory
Chemical Name or Material Sulfuric Acid
Show More Show Less