missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 14% (v/v), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| Concentration | 14% (v/v) |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Percent Purity | 14% V/V |
| Grade | Laboratory |
| Chemical Name or Material | Sulfuric Acid |