missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 10% (v/v), Ricca Chemical

Catalog No. 815016
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
16 gal.
Packaging:
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
815016 500 mL Natural Poly Bottle
815032 1 L Natural Poly Bottle
81505PT 2.5 L Safety-coated Glass Bottle
81501 4 L Natural Poly Bottle
815016G 16 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 815016 Supplier Ricca Chemical Company Supplier No. 815016

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous solution
Quantity 500 mL
Percent Purity 10% V/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10%(v/v)
Chemical Name or Material Sulfuric Acid