missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide sulfurique, 1,00 Normal, Réactifs
Spécifications
Spécifications
| pH | <1 |
| Color | Incolore |
| Boiling Point | 100°C |
| Packaging | Cubitainer |
| Specific Gravity | 1.031 |
| Quantity | 10 L |
| Physical Form | Liquide |
| Grade | Réactif |
| Chemical Name or Material | Sulfuric Acid, 1.00 Normal |