missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 1.00 N, Ricca Chemical

$64.44 - $298.94

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
8300-4
SDS Accessories View Documents
Ricca Chemical Company
83004
120 mL Natural Poly Bottle
Each for $68.22
Only null left
 
830016
SDS Accessories View Documents
Ricca Chemical Company
830016
500 mL Natural Poly Bottle
Each for $64.44
Only null left
 
8300-32
SDS Accessories View Documents
Ricca Chemical Company
830032
1 L Natural Poly Bottle
Each for $79.78
Only null left
 
8300-1
SDS Accessories View Documents
Ricca Chemical Company
83001
4 L Natural Poly Bottle
Each for $199.94
Only null left
 
8300-25
SDS View Documents
Ricca Chemical Company
830025
10 L Cubitainer
Each for $298.94
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O

Specifications

7732-18-5,7664-93-9
1.00N
Natural Poly Bottle
H2O4S
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
Sulfuric Acid
93.0,4.78
97.0,4.97
120 mL
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Laboratory
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'